siRNA Details
| Gene Symbol: Timm8a1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053370.3 | NP_445822.1 | 258 | |||
| GenBank mRNA | AF150082.1 | - | 259 | |||
| BC060552.1 | - | 263 | 6921445 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Timm8a1| Gene Symbol: Timm8a1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053370.3 | NP_445822.1 | 258 | |||
| GenBank mRNA | AF150082.1 | - | 259 | |||
| BC060552.1 | - | 263 | 6921445 | |||