siRNA Details
| Gene Symbol: Il18bp | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053374.2 | NP_445826.2 | 3 | 255 | ||
| XM_008759805.1 | XP_008758027.1 | 3 | 2413 | |||
| GenBank mRNA | AF154569.1 | - | 2 | 66 | ||
| CB809932.1 | - | 3 | 266 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Il18bp| Gene Symbol: Il18bp | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053374.2 | NP_445826.2 | 3 | 255 | ||
| XM_008759805.1 | XP_008758027.1 | 3 | 2413 | |||
| GenBank mRNA | AF154569.1 | - | 2 | 66 | ||
| CB809932.1 | - | 3 | 266 | |||