siRNA Details
| Gene Symbol: Kif3a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053377.1 | NP_445829.1 | 15 | 1955 | ||
| XM_006246347.2 | XP_006246409.1 | 16 | 2065 | |||
| XM_006246351.2 | XP_006246413.1 | 14 | 1983 | |||
| XM_008767716.1 | XP_008765938.1 | 15 | 2143 | |||
| XM_008767717.1 | XP_008765939.1 | 15 | 1992 | |||
| GenBank mRNA | AF155825.1 | - | 7 | 656 | ||

Kif3a
Genomic Map