siRNA Details
| Gene Symbol: Dcx | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053379.3 | NP_445831.3 | 3 | 483 | ||
| XM_006257382.2 | XP_006257444.1 | 3 | 608 | |||
| XM_006257383.2 | XP_006257445.1 | 3 | 701 | |||
| XM_006257384.2 | XP_006257446.1 | 3 | 700 | |||
| XM_006257385.2 | XP_006257447.1 | 3 | 701 | |||
| XM_006257386.2 | XP_006257448.1 | 3 | 701 | |||
| XM_008773448.1 | XP_008771670.1 | 3 | 1170 | |||
| GenBank mRNA | AF155959.2 | - | 2 | 417 | ||

Dcx
Genomic Map