siRNA Details
| Gene Symbol: Slc34a2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053380.2 | NP_445832.1 | 10, 11 | 1239 | ||
| XM_006251032.2 | XP_006251094.1 | 10, 11 | 1328 | |||
| GenBank mRNA | AF157026.1 | - | 9, 10 | 1228 | ||
| AF247725.1 | - | 4, 5 | 567 | |||
| BC070898.1 | - | 10, 11 | 1239 | 7099656 | ||

Slc34a2
Genomic Map