siRNA Details
| Gene Symbol: Puf60 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001191880.1 | NP_001178809.1 | 12 | 1566 | ||
| XM_006241894.2 | XP_006241956.1 | 12 | 1702 | |||
| XM_006241895.2 | XP_006241957.1 | 11 | 1651 | |||
| XM_006241896.2 | XP_006241958.1 | 11 | 1648 | |||
| XM_006241897.2 | XP_006241959.1 | 11 | 1600 | |||
| XM_006241898.2 | XP_006241960.1 | 10 | 1549 | |||
| XM_006241899.2 | XP_006241961.1 | 12 | 1995 | |||
| GenBank mRNA | AF165892.1 | - | 11 | 1555 | ||

Puf60
Genomic Map