siRNA Details
| Gene Symbol: Puf60 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001191880.1 | NP_001178809.1 | 8, 9 | 840 | ||
| XM_006241894.2 | XP_006241956.1 | 8, 9 | 976 | |||
| XM_006241895.2 | XP_006241957.1 | 7, 8 | 925 | |||
| XM_006241896.2 | XP_006241958.1 | 7, 8 | 922 | |||
| XM_006241897.2 | XP_006241959.1 | 7, 8 | 874 | |||
| XM_006241898.2 | XP_006241960.1 | 6, 7 | 823 | |||
| XM_006241899.2 | XP_006241961.1 | 8, 9 | 1269 | |||
| GenBank mRNA | AF165892.1 | - | 7, 8 | 829 | ||

Puf60
Genomic Map