siRNA Details
| Gene Symbol: Cdh8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053393.2 | NP_445845.2 | 11 | 2407 | ||
| XM_008772244.1 | XP_008770466.1 | 10 | 1754 | |||
| GenBank mRNA | AB010436.1 | - | 11 | 2407 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Cdh8| Gene Symbol: Cdh8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053393.2 | NP_445845.2 | 11 | 2407 | ||
| XM_008772244.1 | XP_008770466.1 | 10 | 1754 | |||
| GenBank mRNA | AB010436.1 | - | 11 | 2407 | ||