siRNA Details
| Gene Symbol: Robo2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_008768537.1 | XP_008766759.1 | 15 | 3031 | ||
| XM_008768538.1 | XP_008766760.1 | 15 | 3030 | |||
| XM_008768539.1 | XP_008766761.1 | 15 | 3027 | |||
| GenBank mRNA | AF182037.1 | - | 16 | 2868 | ||

Robo2
Genomic Map