siRNA Details
| Gene Symbol: Smpx | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053395.1 | NP_445847.1 | 5 | 517 | ||
| XM_006256945.2 | XP_006257007.1 | 5 | 486 | |||
| GenBank mRNA | AF364071.1 | - | 5 | 517 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Smpx| Gene Symbol: Smpx | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053395.1 | NP_445847.1 | 5 | 517 | ||
| XM_006256945.2 | XP_006257007.1 | 5 | 486 | |||
| GenBank mRNA | AF364071.1 | - | 5 | 517 | ||