siRNA Details
| Gene Symbol: Maged1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053409.2 | NP_445861.1 | 10 | 1940 | ||
| GenBank mRNA | AF217964.1 | - | 10 | 2076 | ||
| AF274043.1 | - | 8 | 1279 | |||
| AJ133038.1 | - | 8 | 1403 | |||
| BC081756.1 | - | 10 | 1940 | 7134252 | ||

Maged1
Genomic Map