siRNA Details
| Gene Symbol: Ddx1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053414.1 | NP_445866.1 | 8, 9 | 518 | ||
| GenBank mRNA | BC082049.1 | - | 8, 9 | 518 | 7131804 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ddx1| Gene Symbol: Ddx1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053414.1 | NP_445866.1 | 8, 9 | 518 | ||
| GenBank mRNA | BC082049.1 | - | 8, 9 | 518 | 7131804 | |