siRNA Details
| Gene Symbol: Ufd1l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053418.2 | NP_445870.1 | 3, 4 | 274 | ||
| XM_008768904.1 | XP_008767126.1 | 3, 4 | 291 | |||
| GenBank mRNA | BC161818.1 | - | 3, 4 | 285 | 9101868 | |
| CK599711.1 | - | 274 | ||||

Ufd1l
Genomic Map