siRNA Details
| Gene Symbol: Synrg | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053419.1 | NP_445871.1 | 10 | 1319 | ||
| GenBank mRNA | AF169549.1 | - | 3 | 290 | ||
| AF242544.1 | - | 8 | 977 | |||
| EF121977.1 | - | 8 | 782 | |||
| EF121978.1 | - | 8 | 782 | |||
| EF121979.1 | - | 9 | 1016 | |||
| EF121980.1 | - | 9 | 1016 | |||
| EF121981.1 | - | 10 | 1319 | |||
| EF121982.1 | - | 8 | 782 | |||

Synrg
Genomic Map