siRNA Details
| Gene Symbol: Synrg | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053419.1 | NP_445871.1 | 19 | 3755 | ||
| GenBank mRNA | EF121977.1 | - | 17 | 3218 | ||
| EF121978.1 | - | 17 | 3218 | |||
| EF121979.1 | - | 18 | 3452 | |||
| EF121980.1 | - | 18 | 3452 | |||
| EF121981.1 | - | 19 | 3755 | |||
| EF121982.1 | - | 17 | 3218 | |||

Synrg
Genomic Map