siRNA Details
| Gene Symbol: Slc4a4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053424.1 | NP_445876.1 | 10 | 1167 | ||
| XM_006250788.2 | XP_006250850.1 | 10 | 1627 | |||
| XM_006250789.2 | XP_006250851.1 | 10 | 1627 | |||
| XM_006250790.2 | XP_006250852.1 | 11 | 1345 | |||
| XM_006250791.2 | XP_006250853.1 | 10 | 1179 | |||
| GenBank mRNA | AF004017.1 | - | 7 | 1005 | ||
| AF027362.1 | - | 7 | 1037 | |||
| AF124441.1 | - | 10 | 1167 | |||
| AF210250.1 | - | 10 | 1174 | |||
| AF254802.1 | - | 10 | 1167 | |||

Slc4a4
Genomic Map