siRNA Details
| Gene Symbol: Ccs | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053425.1 | NP_445877.1 | 7 | 646 | ||
| GenBank mRNA | AF255305.1 | - | 7 | 646 | ||
| BC158586.1 | - | 7 | 606 | 9099085 | ||
| EU340033.1 | - | 6 | 508 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ccs| Gene Symbol: Ccs | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053425.1 | NP_445877.1 | 7 | 646 | ||
| GenBank mRNA | AF255305.1 | - | 7 | 646 | ||
| BC158586.1 | - | 7 | 606 | 9099085 | ||
| EU340033.1 | - | 6 | 508 | |||