siRNA Details
| Gene Symbol: Trpc4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001083115.1 | NP_001076584.1 | 11 | 2503 | ||
| NM_080396.1 | NP_536321.1 | 10 | 2755 | |||
| XM_006232356.2 | XP_006232418.1 | 11 | 3079 | |||
| XM_008761008.1 | XP_008759230.1 | 10 | 3037 | |||
| GenBank mRNA | AB008889.2 | - | 11 | 2801 | ||
| AF288407.1 | - | 10 | 2771 | |||
| AF288408.1 | - | 11 | 2519 | |||
| AF421363.1 | - | 10 | 2755 | |||
| AF421364.1 | - | 11 | 2503 | |||
| AF421365.1 | - | 11 | 2500 | |||
| AF421366.1 | - | 11 | 2435 | |||
| AF421367.1 | - | 11 | 2450 | |||
| AF421368.1 | - | 11 | 2492 | |||
| FQ228645.1 | - | 11 | 2974 | |||

Trpc4
Genomic Map