siRNA Details
| Gene Symbol: Slco1c1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053441.1 | NP_445893.1 | 5, 6 | 910 | ||
| XM_006237602.1 | XP_006237664.1 | 6, 7 | 1058 | |||
| GenBank mRNA | AF306546.1 | - | 5, 6 | 910 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Slco1c1| Gene Symbol: Slco1c1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053441.1 | NP_445893.1 | 5, 6 | 910 | ||
| XM_006237602.1 | XP_006237664.1 | 6, 7 | 1058 | |||
| GenBank mRNA | AF306546.1 | - | 5, 6 | 910 | ||