siRNA Details
| Gene Symbol: Otof | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001276720.1 | NP_001263649.1 | 28 | 3619 | ||
| XM_006239833.2 | XP_006239895.1 | 29 | 3727 | |||
| XM_008764534.1 | XP_008762756.1 | 28 | 3682 | |||
| XM_008764535.1 | XP_008762757.1 | 28 | 3682 | |||

Otof
Genomic Map