siRNA Details
| Gene Symbol: Tgfb1i1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001191840.1 | NP_001178769.1 | 11 | 1311 | ||
| XM_006230349.2 | XP_006230411.1 | 11 | 2161 | |||
| XM_006230350.2 | XP_006230412.1 | 11 | 1283 | |||
| XM_008759920.1 | XP_008758142.1 | 10 | 2368 | |||
| XM_008759921.1 | XP_008758143.1 | 11 | 1368 | |||
| GenBank mRNA | AF314960.1 | - | 7 | 891 | ||

Tgfb1i1
Genomic Map