siRNA Details
| Gene Symbol: Fads1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053445.2 | NP_445897.2 | 2 | 389 | ||
| XM_006231075.2 | XP_006231137.1 | 2 | 464 | |||
| GenBank mRNA | AB052085.1 | - | 2 | 389 | ||
| AF320509.1 | - | 2 | 418 | |||
| HQ909026.1 | - | 2 | 313 | |||

Fads1
Genomic Map