siRNA Details
| Gene Symbol: Hdac7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_006242364.2 | XP_006242426.1 | 5, 6 | 1560 | ||
| XM_006242365.2 | XP_006242427.1 | 5, 6 | 1560 | |||
| XM_006242366.2 | XP_006242428.1 | 5, 6 | 1560 | |||
| XM_006242367.2 | XP_006242429.1 | 5, 6 | 1560 | |||
| XM_008765842.1 | XP_008764064.1 | 6, 7 | 889 | |||
| XM_008765843.1 | XP_008764065.1 | 5, 6 | 755 | |||
| GenBank mRNA | AF321135.1 | - | 4, 5 | 495 | ||

Hdac7
Genomic Map