siRNA Details
Gene Symbol: Hdac7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006242364.2 | XP_006242426.1 | 4 | 1372 | ||
XM_006242365.2 | XP_006242427.1 | 4 | 1372 | |||
XM_006242366.2 | XP_006242428.1 | 4 | 1372 | |||
XM_006242367.2 | XP_006242429.1 | 4 | 1372 | |||
XM_008765842.1 | XP_008764064.1 | 5 | 701 | |||
XM_008765843.1 | XP_008764065.1 | 4 | 567 | |||
GenBank mRNA | AF321135.1 | - | 3 | 307 |

More Information
Other Species Targeted

Panther Classification:
Gene Ontology Categories:
