siRNA Details
| Gene Symbol: Fgl2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053455.2 | NP_445907.2 | 2 | 987 | ||
| GenBank mRNA | AF323608.2 | - | 2 | 1014 | ||
| BC072502.1 | - | 2 | 936 | |||
| BC085357.1 | - | 2 | 944 | |||
| BC107665.1 | - | 2 | 960 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Fgl2| Gene Symbol: Fgl2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053455.2 | NP_445907.2 | 2 | 987 | ||
| GenBank mRNA | AF323608.2 | - | 2 | 1014 | ||
| BC072502.1 | - | 2 | 936 | |||
| BC085357.1 | - | 2 | 944 | |||
| BC107665.1 | - | 2 | 960 | |||