siRNA Details
| Gene Symbol: Cldn11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053457.2 | NP_445909.2 | 3 | 765 | ||
| GenBank mRNA | AF324043.1 | - | 3 | 570 | ||
| BC070927.1 | - | 3 | 765 | 7096642 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Cldn11| Gene Symbol: Cldn11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053457.2 | NP_445909.2 | 3 | 765 | ||
| GenBank mRNA | AF324043.1 | - | 3 | 570 | ||
| BC070927.1 | - | 3 | 765 | 7096642 | ||