siRNA Details
| Gene Symbol: Srm | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053464.2 | NP_445916.1 | 6, 7 | 825 | ||
| GenBank mRNA | AF337636.1 | - | 6, 7 | 828 | ||
| BC128733.1 | - | 6, 7 | 871 | 8366755 | ||
| Gene Symbol: LOC100912604 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_003749458.2 | XP_003749506.1 | 6, 7 | 822 | ||

Srm
Genomic Map