siRNA Details
| Gene Symbol: Jak1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053466.1 | NP_445918.1 | 9 | 1406 | ||
| XM_006238452.2 | XP_006238514.1 | 9 | 1579 | |||
| XM_006238453.2 | XP_006238515.1 | 9 | 5202 | |||
| GenBank mRNA | AJ000556.1 | - | 5 | 884 | ||

Jak1
Genomic Map