siRNA Details
| Gene Symbol: Sema6b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053471.2 | NP_445923.1 | 5 | 660 | ||
| XM_006244348.2 | XP_006244410.1 | 6 | 1129 | |||
| XM_006244349.2 | XP_006244411.1 | 5 | 482 | |||
| GenBank mRNA | AB000776.1 | - | 5 | 660 | ||

Sema6b
Genomic Map