siRNA Details
| Gene Symbol: Cox4i2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053472.1 | NP_445924.1 | 4 | 318 | ||
| XM_006235280.2 | XP_006235342.1 | 4 | 673 | |||
| GenBank mRNA | AF255347.1 | - | 4 | 318 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Cox4i2| Gene Symbol: Cox4i2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053472.1 | NP_445924.1 | 4 | 318 | ||
| XM_006235280.2 | XP_006235342.1 | 4 | 673 | |||
| GenBank mRNA | AF255347.1 | - | 4 | 318 | ||