siRNA Details
| Gene Symbol: Ppp1r9b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053474.1 | NP_445926.1 | 4, 5 | 2146 | ||
| XM_008768126.1 | XP_008766348.1 | 3, 4 | 1654 | |||
| GenBank mRNA | AF016252.1 | - | 4, 5 | 2146 | ||
| BC160878.1 | - | 4, 5 | 1828 | 7376917 | ||

Ppp1r9b
Genomic Map