siRNA Details
| Gene Symbol: Ppp1r9b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053474.1 | NP_445926.1 | 2, 3 | 1881 | ||
| XM_008768126.1 | XP_008766348.1 | 1, 2 | 1389 | |||
| GenBank mRNA | AF016252.1 | - | 2, 3 | 1881 | ||
| BC160878.1 | - | 2, 3 | 1563 | 7376917 | ||

Ppp1r9b
Genomic Map