siRNA Details
| Gene Symbol: Synj1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053476.2 | NP_445928.2 | 18 | 2350 | ||
| GenBank mRNA | AJ006855.1 | - | 9 | 1305 | ||
| U45479.2 | - | 18 | 2350 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Synj1| Gene Symbol: Synj1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053476.2 | NP_445928.2 | 18 | 2350 | ||
| GenBank mRNA | AJ006855.1 | - | 9 | 1305 | ||
| U45479.2 | - | 18 | 2350 | |||