siRNA Details
| Gene Symbol: Plcb2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053478.1 | NP_445930.1 | 21 | 2370 | ||
| GenBank mRNA | AJ011035.1 | - | 21 | 2370 | ||
| DQ120505.1 | - | 21 | 2296 | |||
| DQ120506.1 | - | 21 | 2296 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Plcb2| Gene Symbol: Plcb2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053478.1 | NP_445930.1 | 21 | 2370 | ||
| GenBank mRNA | AJ011035.1 | - | 21 | 2370 | ||
| DQ120505.1 | - | 21 | 2296 | |||
| DQ120506.1 | - | 21 | 2296 | |||