siRNA Details
| Gene Symbol: Pola2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053480.2 | NP_445932.2 | 2, 3 | 337 | ||
| GenBank mRNA | AJ245648.1 | - | 2, 3 | 206 | ||
| BC097300.1 | - | 2, 3 | 337 | 7457570 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Pola2| Gene Symbol: Pola2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053480.2 | NP_445932.2 | 2, 3 | 337 | ||
| GenBank mRNA | AJ245648.1 | - | 2, 3 | 206 | ||
| BC097300.1 | - | 2, 3 | 337 | 7457570 | ||