siRNA Details
| Gene Symbol: Pik3cb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_006243642.2 | XP_006243704.1 | 20 | 2830 | ||
| XM_008766567.1 | XP_008764789.1 | 15 | 2217 | |||
| GenBank mRNA | AJ012482.1 | - | 20 | 2813 | ||
| Gene Symbol: LOC100910021 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_003750567.3 | XP_003750615.1 | 3 | 689 | ||

Pik3cb
Genomic Map