siRNA Details
| Gene Symbol: Spa17 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053482.1 | NP_445934.1 | 1 | 47 | ||
| XM_006242812.2 | XP_006242874.1 | 2 | 223 | |||
| GenBank mRNA | AJ131888.1 | - | 1 | 47 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Spa17| Gene Symbol: Spa17 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053482.1 | NP_445934.1 | 1 | 47 | ||
| XM_006242812.2 | XP_006242874.1 | 2 | 223 | |||
| GenBank mRNA | AJ131888.1 | - | 1 | 47 | ||