siRNA Details
| Gene Symbol: Col5a2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053488.1 | NP_445940.1 | 53 | 4394 | ||
| GenBank mRNA | AJ224880.1 | - | 10 | 1141 | ||
| FQ222447.1 | - | 53 | 4376 | |||
| FQ229419.1 | - | 3 | 369 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Col5a2| Gene Symbol: Col5a2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053488.1 | NP_445940.1 | 53 | 4394 | ||
| GenBank mRNA | AJ224880.1 | - | 10 | 1141 | ||
| FQ222447.1 | - | 53 | 4376 | |||
| FQ229419.1 | - | 3 | 369 | |||