siRNA Details
| Gene Symbol: Col18a1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053489.2 | NP_445941.2 | 39 | 3643 | ||
| GenBank mRNA | AF189709.1 | - | 2 | 299 | ||
| AJ236873.1 | - | 1 | 150 | |||
| FM035839.1 | - | 2 | 415 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Col18a1| Gene Symbol: Col18a1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053489.2 | NP_445941.2 | 39 | 3643 | ||
| GenBank mRNA | AF189709.1 | - | 2 | 299 | ||
| AJ236873.1 | - | 1 | 150 | |||
| FM035839.1 | - | 2 | 415 | |||