siRNA Details
| Gene Symbol: Xpo1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053490.1 | NP_445942.1 | 8 | 723 | ||
| XM_006251578.2 | XP_006251640.1 | 9 | 1519 | |||
| XM_008770437.1 | XP_008768659.1 | 7 | 1047 | |||
| GenBank mRNA | AB105193.1 | - | 8 | 723 | ||

Xpo1
Genomic Map