siRNA Details
| Gene Symbol: Plg | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053491.2 | NP_445943.1 | 19 | 2358 | ||
| GenBank mRNA | AJ242649.1 | - | 19 | 2361 | ||
| BC091135.1 | - | 19 | 2358 | 7369357 | ||
| FQ210878.1 | - | 19 | 2501 | |||
| FQ218248.1 | - | 6 | 601 | |||

Plg
Genomic Map