siRNA Details
| Gene Symbol: Slc2a8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053494.2 | NP_445946.2 | 8 | 1049 | ||
| GenBank mRNA | AJ245935.1 | - | 8 | 1049 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Slc2a8| Gene Symbol: Slc2a8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053494.2 | NP_445946.2 | 8 | 1049 | ||
| GenBank mRNA | AJ245935.1 | - | 8 | 1049 | ||