siRNA Details
| Gene Symbol: Slc2a8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053494.2 | NP_445946.2 | 9 | 1313 | ||
| XM_008761779.1 | XP_008760001.1 | 8 | 1180 | |||
| GenBank mRNA | AJ245935.1 | - | 9 | 1313 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Slc2a8| Gene Symbol: Slc2a8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053494.2 | NP_445946.2 | 9 | 1313 | ||
| XM_008761779.1 | XP_008760001.1 | 8 | 1180 | |||
| GenBank mRNA | AJ245935.1 | - | 9 | 1313 | ||