siRNA Details
| Gene Symbol: Entpd6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053498.1 | NP_445950.1 | 4, 5 | 648 | ||
| XM_006235218.2 | XP_006235280.1 | 4, 5 | 743 | |||
| GenBank mRNA | AJ277748.1 | - | 4, 5 | 648 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Entpd6| Gene Symbol: Entpd6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053498.1 | NP_445950.1 | 4, 5 | 648 | ||
| XM_006235218.2 | XP_006235280.1 | 4, 5 | 743 | |||
| GenBank mRNA | AJ277748.1 | - | 4, 5 | 648 | ||