siRNA Details
| Gene Symbol: Ajuba | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053503.1 | NP_445955.1 | 1 | 364 | ||
| GenBank mRNA | BC085802.1 | - | 1 | 364 | 7110970 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ajuba| Gene Symbol: Ajuba | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053503.1 | NP_445955.1 | 1 | 364 | ||
| GenBank mRNA | BC085802.1 | - | 1 | 364 | 7110970 | |