siRNA Details
| Gene Symbol: Prss12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053504.1 | NP_445956.1 | 8 | 1738 | ||
| GenBank mRNA | AJ311671.2 | - | 8 | 1738 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Prss12| Gene Symbol: Prss12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053504.1 | NP_445956.1 | 8 | 1738 | ||
| GenBank mRNA | AJ311671.2 | - | 8 | 1738 | ||