siRNA Details
| Gene Symbol: Nme3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053507.1 | NP_445959.1 | 4 | 369 | ||
| XM_008767598.1 | XP_008765820.1 | 4 | 481 | |||
| GenBank mRNA | AY017337.1 | - | 4 | 369 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Nme3| Gene Symbol: Nme3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053507.1 | NP_445959.1 | 4 | 369 | ||
| XM_008767598.1 | XP_008765820.1 | 4 | 481 | |||
| GenBank mRNA | AY017337.1 | - | 4 | 369 | ||