siRNA Details
| Gene Symbol: Fbxo2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053511.1 | NP_445963.1 | 6 | 1019 | ||
| XM_008764298.1 | XP_008762520.1 | 7 | 1222 | |||
| GenBank mRNA | AF098301.1 | - | 6 | 1019 | ||
| FQ214021.1 | - | 6 | 1012 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Fbxo2| Gene Symbol: Fbxo2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053511.1 | NP_445963.1 | 6 | 1019 | ||
| XM_008764298.1 | XP_008762520.1 | 7 | 1222 | |||
| GenBank mRNA | AF098301.1 | - | 6 | 1019 | ||
| FQ214021.1 | - | 6 | 1012 | |||