siRNA Details
| Gene Symbol: Prdx4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053512.2 | NP_445964.1 | 4 | 599 | ||
| XM_006256951.2 | XP_006257013.1 | 4 | 764 | |||
| GenBank mRNA | AF106945.1 | - | 4 | 600 | ||
| BC059122.1 | - | 4 | 683 | 6918063 | ||
| FQ219258.1 | - | 4 | 647 | |||
| FQ220216.1 | - | 4 | 653 | |||
| FQ220257.1 | - | 4 | 647 | |||
| FQ220425.1 | - | 4 | 648 | |||
| FQ220916.1 | - | 4 | 648 | |||
| FQ223235.1 | - | 4 | 643 | |||
| FQ228535.1 | - | 4 | 648 | |||

Prdx4
Genomic Map