siRNA Details
| Gene Symbol: Pla1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_138882.1 | NP_620237.1 | 10 | 1169 | ||
| XM_006248377.2 | XP_006248439.1 | 10 | 1582 | |||
| GenBank mRNA | BC078727.1 | - | 10 | 1169 | 7124402 | |
| D88666.1 | - | 10 | 1169 | |||
| FQ210605.1 | - | 3 | 162 | |||

Pla1a
Genomic Map